Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium sputtering target, 76.2mm dia x 1.59mm thick

$1,628.81 - $1,628.81

Chemical Identifiers

Chemical Name or Material Gadolinium sputtering target
CAS 7440-54-2
Name Note 76.2mm dia. x 1.59mm thick
MDL Number MFCD00011022
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA46061KS

 
alfa aesar™
46061KS
1 ea. Each for $1,628.81
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Gadolinium sputtering target
TSCA TSCA
Name Note 76.2mm dia. x 1.59mm thick
MDL Number MFCD00011022
CAS 7440-54-2