missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium sputtering target, 50.8mm dia x 1.59mm thick

$551.07 - $551.07

Chemical Identifiers

CAS 7440-54-2
MDL Number MFCD00011022
Chemical Name or Material Gadolinium sputtering target, 50.8mm dia. x 1.59mm thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45621KS SDS Alfa Aesar™
45621KS
1 ea. Each for $551.07
Specifications Description & Specifications

Chemical Identifiers

7440-54-2
MFCD00011022
Gadolinium sputtering target, 50.8mm dia. x 1.59mm thick

Specifications

1 ea.
7440-54-2
MFCD00011022
Gadolinium sputtering target, 50.8mm dia. x 1.59mm thick

Safety and Handling

TSCA : TSCA