Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium rod, 12.7mm (0.5 in.) dia., 99.9% (metals basis excluding Ta)

Chemical Identifiers

Chemical Name or Material Gadolinium rod
CAS 7440-54-2
Name Note 12.7mm (0.5 in.) dia.
MDL Number MFCD00011022
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40290DH

  SDS
alfa aesar™
40290DH
25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40290HF

  SDS
alfa aesar™
40290HF
10mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Cast Rod, dia. is approximate
Quantity 25mm
Packaging Mylar under Argon
Name Note 12.7mm (0.5 in.) dia.
MDL Number MFCD00011022
Chemical Name or Material Gadolinium rod
TSCA TSCA
Assay 99.9%
Weight ≈0.97g/mm
Storage Note 1
  • Packaged under argon
Assay Percent Notes (metals basis excluding Ta)
CAS 7440-54-2

  • Packaged under argon

Safety and Handling

  • Packaged under argon