missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium powder, -200 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4591506

 View more versions of this product

Catalog No. AA4591506



Description

Description

  • Packaged under Argon
Specifications

Specifications

Gadolinium powder
Air & Moisture Sensitive
5g
Odorless
(REO)
MFCD00011022
UIWYJDYFSGRHKR-UHFFFAOYSA-N
gadolinium
23982
Powder
−200 Mesh
Silver
7440-54-2
99.9%
Gd
UN3089
[Gd]
157.25
CHEBI:33375

Chemical Identifiers

7440-54-2
157.25
UIWYJDYFSGRHKR-UHFFFAOYSA-N
CHEBI:33375
[Gd]
Gd
MFCD00011022
23982
gadolinium
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378b-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : LW3850000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents