Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium pieces, 20mm (0.79 in.) & down, 99.9% (REO)

$125.07 - $125.07

Chemical Identifiers

Chemical Name or Material Gadolinium pieces
CAS 7440-54-2
MDL Number MFCD00011022
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA0011809

  SDS
alfa aesar™
0011809*
10g Each for $125.07
Description & Specifications

Specifications

Quantity 10g
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Gadolinium pieces
Packaging Glass bottle
TSCA TSCA
Physical Form Pieces
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011022
CAS 7440-54-2
UN Number UN3178
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Particle Size 20mm (0.79 in.) & down
RTECS Number LW3850000
Color Silver

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

  • Packaged under argon