missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) oxide, nanopowder, 99.99+% (REO)

Manufacturer:  Alfa Aesar™ 4402036

 View more versions of this product

Catalog No. AA4402036



Specifications

Specifications

Gadolinium (III) oxide
(REO)
MFCD00011026
digadolinium 3+ ion trioxidandiide
CMIHHWBVHJVIGI-UHFFFAOYSA-N
gadolinium(3+);oxygen(2-)
159427
20-40nm Powder
7.407 g/mL
2330°C
12064-62-9
Gd2O3
14,4326
Soluble in acid. Insoluble in water.
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
362.497
362.5
≥99.99%
20 to 40nm
500g

Chemical Identifiers

12064-62-9
362.497
CMIHHWBVHJVIGI-UHFFFAOYSA-N
159427
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
Gd2O3
MFCD00011026
digadolinium 3+ ion trioxidandiide
gadolinium(3+);oxygen(2-)
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 235-060-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents