missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium(III) oxide, 99.999% (REO), Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1128914

 View more versions of this product

Catalog No. AA1128914



Specifications

Specifications

Gadolinium(III) oxide
99.999% (REO)
MFCD00011026
gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
362.497
362.5
7.407g/cm3
25g
12064-62-9
Gd2O3
White powder or granules
CMIHHWBVHJVIGI-UHFFFAOYSA-N
gadolinium(3+);oxygen(2-)
159427
Powder or Granular
White

Chemical Identifiers

12064-62-9
362.497
CMIHHWBVHJVIGI-UHFFFAOYSA-N
159427
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
Gd2O3
MFCD00011026
gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
gadolinium(3+);oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 235-060-9

Documents