Please login to your online account to display your discounted pricing

Gadolinium(III) oxide, 99.9% (trace metal basis), ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC315511000

 View more versions of this product

Catalog No. AC315511000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 362.5
Quantity 100g
Color White
Purity Grade For trace metal basis analysis
Merck Index 10, 4200
Chemical Name or Material Gadolinium(III) oxide
Packaging Glass bottle
Assay Percent Range 99.90%
Molecular Formula Gd2O3
Linear Formula GdO3
MDL Number MFCD00011026
CAS 12064-62-9
Total Impurity 0.1% max.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Gadolinium(III) oxide
CAS 12064-62-9
Purity Grade For trace metal basis analysis
Assay Percent Range 99.90%
Molecular Formula Gd2O3
Linear Formula GdO3
Formula Weight 362.5
MDL Number MFCD00011026
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents