Please login to your online account to display your discounted pricing

Gadolinium(III) oxide, 99.9% (REO), Alfa Aesar™, REacton™

$67.19 - $67.19

Chemical Identifiers

Chemical Name or Material Gadolinium(III) oxide
CAS 12064-62-9
Physical Form Powder or Granular
Assay Percent Range 99.9% (REO)
Molecular Formula Gd2O3
Formula Weight 362.5
MDL Number MFCD00011026
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1129114

  SDS
alfa aesar
1129114
25g Each for $67.19

AA1129136

  SDS
alfa aesar
1129136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1129122

  SDS
alfa aesar
1129122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gadolinium(III) oxide
CAS 12064-62-9
EINECS Number 235-060-9
Physical Form Powder or Granular
Assay Percent Range 99.9% (REO)
Molecular Formula Gd2O3
Appearance White powder or granules
Formula Weight 362.5
MDL Number MFCD00011026
Density 7.407g/cm3
Color White
Quantity 25g