missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium(III) oxide, 99.9% (REO), Alfa Aesar™, REacton™

Manufacturer:  Thermo Fisher Scientific 1129122

 View more versions of this product

Catalog No. AA1129122



Specifications

Specifications

Gadolinium(III) oxide
99.9% (REO)
MFCD00011026
digadolinium 3+ ion trioxidandiide
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
362.497
362.5
7.407g/cm3
100g
12064-62-9
Gd2O3
White powder or granules
CMIHHWBVHJVIGI-UHFFFAOYSA-N
gadolinium(3+);oxygen(2-)
159427
Powder or Granular
White

Chemical Identifiers

12064-62-9
362.497
CMIHHWBVHJVIGI-UHFFFAOYSA-N
159427
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
Gd2O3
MFCD00011026
digadolinium 3+ ion trioxidandiide
gadolinium(3+);oxygen(2-)
Safety and Handling

Safety and Handling

EINECSNumber : 235-060-9

Documents