Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) oxalate hydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Gadolinium(III) oxalate hydrate
CAS 100655-00-3
Linear Formula Gd2(C2O4)3·xH2O
Formula Weight 578.56 (Anhydrous)
MDL Number MFCD00150145
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1530822

 
alfa aesar™
1530822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1530814

 
alfa aesar™
1530814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 578.56 (Anhydrous)
Melting Point 110°C (in -6H2O)
Chemical Name or Material Gadolinium(III) oxalate hydrate
TSCA TSCA
Physical Form Powder
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
Linear Formula Gd2(C2O4)3·xH2O
MDL Number MFCD00150145
CAS 100655-00-3
UN Number UN3288
Color White
Quantity 100g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.