missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) oxalate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1530822

 View more versions of this product

Catalog No. AA1530822



Specifications

Specifications

Gadolinium(III) oxalate hydrate
(REO)
Gd2(C2O4)3·xH2O
UN3288
FEIDMMDEDUAFJQ-UHFFFAOYSA-N
gadolinium;oxalic acid;hydrate
92020445
Powder
Hygroscopic
110°C (in -6H2O)
100655-00-3
C6H8Gd2O13
MFCD00150145
oxalic acid-gadolinium-water 3/2/1
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Gd].[Gd]
602.617
578.56 (Anhydrous)
99.9%
White
100g

Chemical Identifiers

100655-00-3
602.617
FEIDMMDEDUAFJQ-UHFFFAOYSA-N
92020445
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.[Gd].[Gd]
C6H8Gd2O13
MFCD00150145
oxalic acid-gadolinium-water 3/2/1
gadolinium;oxalic acid;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents