missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gadolinium(III) nitride, 99.5% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04670903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gadolinium (III) nitride | |
25764-15-2 | |
1 g | |
GdN | |
gadolinium nitride, gadolinium iii nitride, gadoliniumylidyneamine | |
N#[Gd] | |
171.26 | |
171.26 |
Black | |
Powder | |
(REO) | |
MFCD00049943 | |
FLATXDRVRRDFBZ-UHFFFAOYSA-N | |
azanylidynegadolinium | |
117632 | |
99.5% |
Chemical Identifiers
25764-15-2 | |
171.26 | |
FLATXDRVRRDFBZ-UHFFFAOYSA-N | |
117632 | |
N#[Gd] |
GdN | |
MFCD00049943 | |
gadolinium nitride, gadolinium iii nitride, gadoliniumylidyneamine | |
azanylidynegadolinium |
Safety and Handling
EINECSNumber : 247-250-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only