missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) nitride, 99.5% (REO)

Manufacturer:  Alfa Aesar™ 4670906

 View more versions of this product

Catalog No. AA4670906



Specifications

Specifications

Gadolinium (III) nitride
99.5%
GdN
FLATXDRVRRDFBZ-UHFFFAOYSA-N
azanylidynegadolinium
117632
Powder
Black
25764-15-2
(REO)
MFCD00049943
N#[Gd]
171.257
171.26
−60 Mesh
5g

Chemical Identifiers

25764-15-2
171.257
FLATXDRVRRDFBZ-UHFFFAOYSA-N
azanylidynegadolinium
GdN
MFCD00049943
117632
N#[Gd]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents