missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium(III) nitrate pentahydrate, 99.999%, (trace metal basis), ACROS Organics™

$103.95 - $103.95

Chemical Identifiers

CAS 52788-53-1
Assay Percent Range 99.999%
Molecular Formula GdN3O9·5H2O
Linear Formula Gd(NO3)3·5H2O
Formula Weight 433.33
MDL Number MFCD00149729
Chemical Name or Material Gadolinium(III) nitrate pentahydrate
Grade Trace Metal Basis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC202800100 SDS Thermo Fisher Scientific
202800100
Glass bottle 10g Each for $103.95
AC202800500 SDS Thermo Fisher Scientific
AC202800500
Glass bottle 50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For ultra trace analysis
Description & Specifications

Chemical Identifiers

52788-53-1
GdN3O9·5H2O
433.33
Gadolinium(III) nitrate pentahydrate
99.999%
Gd(NO3)3·5H2O
MFCD00149729
Trace Metal Basis

Specifications

52788-53-1
99.999%
Gd(NO3)3·5H2O
MFCD00149729
Glass bottle
White
Gadolinium(III) nitrate pentahydrate
Granules
GdN3O9·5H2O
433.33
14, 4326
0.001% max.
10g
Trace Metal Basis

Safety and Handling

GHS H Statement:
Causes serious eye damage.
May intensify fire; oxidizer.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger