Please login to your online account to display your discounted pricing

Gadolinium(III) nitrate pentahydrate, 99.999%, (trace metal basis), ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC202800500

 View more versions of this product

Catalog No. AC202800500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Formula Weight 433.33
Physical Form Granules
Quantity 50g
Color White
Merck Index 14, 4326
Packaging Glass bottle
Linear Formula Gd(NO3)3·5H2O
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May intensify fire; oxidizer.
CAS 52788-53-1
MDL Number MFCD00149729
Chemical Name or Material Gadolinium(III) nitrate pentahydrate
Molecular Formula GdN3O9·5H2O
Assay Percent Range 99.999%
Total Trace Metal Impurities 0.001% max.

Specifications

Chemical Name or Material Gadolinium(III) nitrate pentahydrate
CAS 52788-53-1
Purity Grade Trace Metal Basis
Assay Percent Range 99.999%
Molecular Formula GdN3O9·5H2O
Linear Formula Gd(NO3)3·5H2O
Formula Weight 433.33
MDL Number MFCD00149729
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May intensify fire; oxidizer.

SDS & Safety Documents