Please login to your online account to display your discounted pricing

Gadolinium(III) nitrate pentahydrate, 99.999%, (trace metal basis), ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  202800100

 View more versions of this product

Catalog No. AC202800100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 433.33
Purity Grade Trace Metal Basis
Physical Form Granules
Quantity 10g
Color White
Merck Index 14, 4326
Chemical Name or Material Gadolinium(III) nitrate pentahydrate
Linear Formula Gd(NO3)3·5H2O
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 52788-53-1
Assay Percent Range 99.999%
Molecular Formula GdN3O9·5H2O
MDL Number MFCD00149729
Packaging Glass bottle
Total Trace Metal Impurities 0.001% max.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Gadolinium(III) nitrate pentahydrate
CAS 52788-53-1
Purity Grade Trace Metal Basis
Assay Percent Range 99.999%
Molecular Formula GdN3O9·5H2O
Linear Formula Gd(NO3)3·5H2O
Formula Weight 433.33
MDL Number MFCD00149729
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May intensify fire; oxidizer.

SDS & Safety Documents