Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) nitrate hydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Gadolinium(III) nitrate hydrate
CAS 94219-55-3
Purity Grade Reacton™
Formula Weight 343.26 (Anhydrous)
MDL Number MFCD00011023
UN Number UN1477
Merck Index 14,4326
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1128406

 
alfa aesar™
1128406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1128414

 
alfa aesar™
1128414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1128422

 
alfa aesar™
1128422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 343.26 (Anhydrous)
Chemical Name or Material Gadolinium(III) nitrate hydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Crystalline Aggregates
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00011023
CAS 94219-55-3
UN Number UN1477
Merck Index 14,4326
Color White
Melting Point 92°C
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.406g/mL
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
RTECS Number LW4550000
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.