Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) isopropoxide, 99.9% (metals basis), 5% w/v in toluene/isopropanol (3:2)

Precursor for Gd2O3 films using Sol-Gel process

Chemical Identifiers

Chemical Name or Material Gadolinium(III) isopropoxide
CAS 14532-05-9
Linear Formula Gd[OCH(CH3)2]3
MDL Number MFCD00144381
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42277AE

  SDS
alfa aesar™
42277AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42277AC

  SDS
alfa aesar™
42277AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Gadolinium(III) isopropoxide
Concentration 5% w/v in toluene/isopropanol (3:2)
Sensitivity Moisture Sensitive
Quantity 100mL
CAS 14532-05-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P301+P330+P331-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Physical Form Liquid
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula Gd[OCH(CH3)2]3
MDL Number MFCD00144381
UN Number UN1993
Odor Aromatic
Color Pale yellow
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P301+P330+P331-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H304-H361-H373-H315-H319-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.