Please login to your online account to display your discounted pricing

Gadolinium(III) fluoride 99.9%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Gadolinium(III) fluoride
Assay ≥99.85% (ex fluor)
Assay Percent Range 99.9%
Molecular Formula F3 Gd
Linear Formula GdF3
Formula Weight 214.24
MDL Number MFCD00016113
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383250050

  SDS
thermo fisher scientific
AC383250050
Plastic bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.85% (ex fluor)
Physical Form Powder
Formula Weight 214.24
Quantity 5g
Color White
Packaging Plastic bottle
Chemical Name or Material Gadolinium(III) fluoride
Molecular Formula F3 Gd
Assay Percent Range 99.9%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with acids liberates toxic gas.
MDL Number MFCD00016113
Linear Formula GdF3
CAS 13765-26-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with acids liberates toxic gas.