Please login to your online account to display your discounted pricing

Gadolinium (III) Chloride, Ultra Dry, 99.99%, Alfa Aesar

$171.77 - $171.77

Chemical Identifiers

Chemical Name or Material Gadolinium(III) chloride
CAS 10138-52-0
Physical Form Ultra Dry Powder
Assay Percent Range 99.99% (REO)
Molecular Formula GdCl3
Formula Weight 263.61
MDL Number MFCD00011024
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3573606

  SDS
alfa aesar
3573606
5g Each for $171.77

AA3573614

  SDS
alfa aesar
3573614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Ultra Dry Powder
Formula Weight 263.61
Quantity 5g
Color White
Density 4.52g/cm3
Chemical Name or Material Gadolinium(III) chloride
EINECS Number 233-386-6
Molecular Formula GdCl3
Appearance White powder
MDL Number MFCD00011024
CAS 10138-52-0
Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
Assay Percent Range 99.99% (REO)
Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.