Please login to your online account to display your discounted pricing

Gadolinium(III) chloride hexahydrate, White crystalline aggregates, REacton™, 99.9% (REO), Alfa Aesar™

$34.78 - $272.45

Chemical Identifiers

Chemical Name or Material Gadolinium(III) chloride hexahydrate
Name Note Rare Earth Oxides Content
CAS 13450-84-5
Physical Form Crystalline Aggregates
Assay Percent Range 99.9% (REO)
Molecular Formula GdCl3·6H2O
Formula Weight 371.70 (263.61anhydrous)
MDL Number MFCD00149732
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1128614

  SDS
alfa aesar
1128614
25g Each for $96.82

AA11286A1

  SDS
alfa aesar
11286A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1128606

  SDS
alfa aesar
1128606
5g Each for $34.78

AA1128622

  SDS
alfa aesar
1128622
100g Each for $272.45
Description & Specifications

Specifications

Physical Form Crystalline Aggregates
Formula Weight 371.70 (263.61 Anhydrous)
Quantity 25g
Color White
Density 2.424g/cm3
Chemical Name or Material Gadolinium(III) chloride hexahydrate
EINECS Number 233-386-6
Name Note Rare Earth Oxides Content
Assay Percent Range 99.9% (REO)
Molecular Formula GdCl3·6H2O
MDL Number MFCD00149732
CAS 13450-84-5
Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.