Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Gadolinium(III) chloride hexahydrate
CAS 13450-84-5
Purity Grade Reacton™
Formula Weight 371.70 (263.61 Anhydrous)
MDL Number MFCD00149732
Merck Index 14,4326
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1128706

  SDS
alfa aesar™
1128706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1128714

  SDS
alfa aesar™
1128714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1128722

  SDS
alfa aesar™
1128722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 371.70 (263.61 Anhydrous)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Sensitivity Hygroscopic
Purity Grade Reacton™
Health Hazard 1 Warning
Quantity 5g
Chemical Name or Material Gadolinium(III) chloride hexahydrate
Physical Form Crystalline Aggregates
CAS 13450-84-5
TSCA TSCA
RTECS Number LW4050000
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Odor Characteristic
MDL Number MFCD00149732
Density 2.424g/mL
Merck Index 14,4326
Color White
Assay 99.99%
Assay Percent Notes (REO)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.