missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1320406

 View more versions of this product

Catalog No. AA1320406



Specifications

Specifications

Gadolinium(III) 2,4-pentanedionate hydrate
(REO)
MFCD00013493
NJOCOKMBKFILAF-KJVLTGTBSA-K
gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
131675701
Powder
Reacton™
5g
64438-54-6
C15H23GdO7
gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Gd+3]
472.592
454.58 (Anhydrous)
99.9%
Hygroscopic

Chemical Identifiers

64438-54-6
472.592
NJOCOKMBKFILAF-KJVLTGTBSA-K
131675701
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Gd+3]
C15H23GdO7
MFCD00013493
gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents