missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium ICP Standard, 1mL = 1mg Gd (1,000ppm Gd), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPGD1KN500

 View more versions of this product

Catalog No. PGD1KN-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Gadolinium ICP Standard
93.0,4.56,0.11
Gd2O3
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
362.497
Liquid
1,000ppm Gd
Colorless
12064-62-9,7697-37-2,7732-18-5
97.0,4.74,0.11
CMIHHWBVHJVIGI-UHFFFAOYSA-N
gadolinium(3+);oxygen(2-)
159427
Poly natural
1 mL = 1 mg Gd
500mL

Chemical Identifiers

12064-62-9
362.497
159427
[O-2].[O-2].[O-2].[Gd+3].[Gd+3]
Gd2O3
CMIHHWBVHJVIGI-UHFFFAOYSA-N
gadolinium(3+);oxygen(2-)
Documents