missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12396FF

 View more versions of this product

Catalog No. AA12396FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Gadolinium foil
25 x 25 mm
7440-54-2
Gd
UN3178
Insoluble in water. Soluble in dilute acid.
[Gd]
157.25
CHEBI:33375
99.9%
Silver
≈0.49g/25x25mm
0.1mm (0.004 in.) thick
(REO)
MFCD00011022
atom
UIWYJDYFSGRHKR-UHFFFAOYSA-N
gadolinium
23982
Foil
Air and moisture sensitive

Chemical Identifiers

7440-54-2
157.25
UIWYJDYFSGRHKR-UHFFFAOYSA-N
23982
gadolinium
Gd
MFCD00011022
atom
CHEBI:33375
[Gd]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-162-2

RTECSNumber : LW3850000

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

Documents