Learn More
Alfa Aesar™ Gadolinium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)
Manufacturer: Alfa Aesar™ 12396FI
Description
- Packaged under argon
Specifications
Gadolinium foil | |
≈0.49g/25x25mm | |
50 x 50mm | |
7440-54-2 | |
Gd | |
UN3178 | |
UIWYJDYFSGRHKR-UHFFFAOYSA-N | |
gadolinium | |
23982 | |
Foil |
Air & Moisture Sensitive | |
Silver | |
0.1mm (0.004 in.) thick | |
(REO) | |
MFCD00011022 | |
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds | |
[Gd] | |
157.25 | |
CHEBI:33375 | |
99.9% |
Chemical Identifiers
7440-54-2 | |
157.25 | |
UIWYJDYFSGRHKR-UHFFFAOYSA-N | |
23982 | |
gadolinium |
Gd | |
MFCD00011022 | |
atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds | |
CHEBI:33375 | |
[Gd] |
Safety and Handling
GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
RTECSNumber : LW3850000
TSCA : TSCA
storageNote1 :
- Packaged under argon