missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium foil. 0.075mm (0.003 in.) thick, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 43307FF

 View more versions of this product

Catalog No. AA43307FF



Description

Description

  • Packaged under Argon
Specifications

Specifications

Gadolinium foil. 0.075mm (0.003in) thick
Gray
7440-54-2
Gd
UN3178
UIWYJDYFSGRHKR-UHFFFAOYSA-N
gadolinium
23982
Foil
Reacton™
≈0.36g/25x25mm
25 x 25mm
(REO)
MFCD00011022
atom
[Gd]
157.25
CHEBI:33375
99.9%

Chemical Identifiers

7440-54-2
157.25
UIWYJDYFSGRHKR-UHFFFAOYSA-N
23982
gadolinium
Gd
MFCD00011022
atom
CHEBI:33375
[Gd]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

RTECSNumber : LW3850000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Documents