Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium fluoride, anhydrous, REacton™, elec. gd, low oxy. 99.99%

Chemical Identifiers

Chemical Name or Material Gadolinium fluoride
CAS 13765-26-9
Name Note Anhydrous, Low oxygen
Purity Grade Reacton™
Linear Formula GdF3
Formula Weight 214.25g/mol
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1129206

 
alfa aesar™
11292-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Quantity 5g
Quantity 5g
Chemical Name or Material Gadolinium fluoride
Name Note Anhydrous, Low oxygen
CAS 13765-26-9
Purity Grade Reacton™
Physical Form Crystalline lump in ampoules
Assay 99.99%
Linear Formula GdF3
Formula Weight 214.25g/mol
UN Number UN3288
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.