Please login to your online account to display your discounted pricing

Alfa Aesar™ Gadolinium fluoride, anhydrous, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Gadolinium fluoride
CAS 13765-26-9
Name Note Anhydrous
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3310614

 
alfa aesar™
33106-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 7.1g/mL
Odor Odorless
Quantity 25g
Color White
Melting Point 1231°C
Quantity 25g
Chemical Name or Material Gadolinium fluoride
Name Note Anhydrous
CAS 13765-26-9
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (REO)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.