Please login to your online account to display your discounted pricing
Thermo Scientific™ GABLE COVER PLEXIGLASS SHKNBTH
Prevent cross-contamination by diverting condensation to sides for return to bath medium.
$1,409.91 - $1,409.91
Products
Catalog Number Mfr. No. Material For Use With Price Quantity    
15-453-202D
thermo sci baths
118111
Polycarbonate Models SWB1122, RSWB3222 Each for $1,409.91
Description & Specifications
Prevent cross-contamination by diverting condensation to sides for return to bath medium with Thermo Scientific™ Gable Bath Covers.
  • Reduce moisture loss and water accumulation on the bench
  • Stainless steel or semi-transparent polycarbonate
  • Can be attached to side of bath
  • With handle