missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

G418 Sulfate (White to Off-white Powder), Fisher BioReagents

Manufacturer:  Fisher Scientific Company FLBP6731

 View more versions of this product

Catalog No. BP673-1


 

Specifications Description & Specifications

Specifications

G418 Sulfate
108321-42-2
Solid
C20H40N4O10 · 2 H2SO4
Amber Glass
> 98%
10% Max.
1g
< 0.015 at 280nm
1.0 max.
> 98 %
692.7
> 730μg/mg
Pass Test
White

Chemical Identifiers

108321-42-2
> 98 %
692.7
Solid
C20H40N4O10 · 2 H2SO4
G418 Sulfate

Safety and Handling

SDS & Safety Documents

DANGER!

Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe vapors/dust. Do not ingest. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:1
Instability:0

Recommended Storage : -20°C