missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Gα i1 G-Protein Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ Gαi1 G-Protein Ab-3, Mouse Monoclonal Antibody.

$273.86 - $1,230.82

Specifications

Antigen Galphai1 G-Protein Ab-3
Clone R4.5
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS243P Thermo Scientific™ Lab Vision™
MS243P
200μg/mL 1mL Each for $1,132.96
MS243P0 Thermo Scientific™ Lab Vision™
MS243P0
200μg/mL 100μL Each for $273.86
MS243P1 Thermo Scientific™ Lab Vision™
MS243P1
200μg/mL 500μL Each for $743.69
MS243P1ABX Thermo Scientific™ Lab Vision™
MS243P1ABX
1mg/mL 100μL Each for $832.69
MS243PABX Thermo Scientific™ Lab Vision™
MS243PABX
1mg/mL 200μL Each for $1,230.82
Specifications

Description

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ Gαi1 G-Protein Ab-3, Mouse Monoclonal Antibody.

GTP-binding proteins (G-proteins) play a critical role in signal transduction by coupling receptors to effectors. The intracellular consequences of G-protein activation include second messenger generation, protein phosphorylation, ion channel activation, gene induction, cell growth, and differentiation. Gi belongs to the pertussis toxin sensitive G-proteins. Gi consists of three different subtypes of α-subunits (αi1, αi2 and αi3)

Host Species: Mouse

Clone: R4.5

Isotype: IgG2b

Species Reactivity: Human, Rat, Mouse, Cow, and Guinea Pig. Others not known.

Epitope: Not determined

Immunogen: Partially purified rat brain Gαi1 protein

Molecular Weight: 39-42 kDa

Positive Control: IMR-5 cells

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Galphai1 G-Protein Ab-3
Western Blot
Unlabeled
Murine
Cytoskeletal Protein
Partially purified rat brain Galphai1 protein
R4.5
Monoclonal
Concentrated
RUO
Primary
IgG2b