missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Gα i1 G-Protein Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ Gαi1 G-Protein Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS243P1ABX

 View more versions of this product

Catalog No. MS243P1ABX


 

Specifications

Description

Ensure accurate, reproducible results in western blotting experiments with Thermo Scientific™ Gαi1 G-Protein Ab-3, Mouse Monoclonal Antibody.

GTP-binding proteins (G-proteins) play a critical role in signal transduction by coupling receptors to effectors. The intracellular consequences of G-protein activation include second messenger generation, protein phosphorylation, ion channel activation, gene induction, cell growth, and differentiation. Gi belongs to the pertussis toxin sensitive G-proteins. Gi consists of three different subtypes of α-subunits (αi1, αi2 and αi3)

Host Species: Mouse

Clone: R4.5

Isotype: IgG2b

Species Reactivity: Human, Rat, Mouse, Cow, and Guinea Pig. Others not known.

Epitope: Not determined

Immunogen: Partially purified rat brain Gαi1 protein

Molecular Weight: 39-42 kDa

Positive Control: IMR-5 cells

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Galphai1 G-Protein Ab-3
Monoclonal
1mg/mL
Concentrated
Partially purified rat brain Galphai1 protein
100μL
Cytoskeletal Protein
Monoclonal
Western Blot
R4.5
Unlabeled
Murine
IgG2b
RUO
Primary
Bovine,Guinea Pig,Human,Murine,Rat