missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

G-CSF Rat anti-Mouse, eFluor 660, Clone: 9B4CSF, eBioscience™

Rat Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen G-CSF
Clone 9B4CSF
Host Species Rat
Gene Alias Granulocyte colony-stimulating factor, CSF3
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124454 Life Technologies
LS50735380
25μg Each for $157.08
501124455 Life Technologies
LS50735382
100μg Each for $394.68
Specifications

Description

This 9B4CSF monoclonal antibody reacts with mouse G-CSF (Granulocyte Colony-Stimulating Factor), a growth factor that stimulates the proliferation, differentiation, and mobilization of hematopoetic stem cells. It is expressed by monocytes, macrophages, and bone marrow stromal cells, and can also be induced in fibroblasts by IL-17A. Unlike GM-CSF and IL-3, which can stimulate cells of multiple lineages, G-CSF activity is limited to neutrophilic granulocytes. G-CSF is essential to the maintenance of neutrophil counts during homeostasis, and low basal levels of the protein are detectable in the serum of healthy individuals. Circulating levels become elevated rapidly upon infection, as G-CSF is also important for the activation and mobilization of mature neutrophils during the innate immune response. G-CSF therapy is commonly used to treat neutropenia following bone marrow transplant or chemotherapy.

This 9B4CSF antibody has been demonstrated to cross-react with human G-CSF.
Description & Specifications

Specifications

G-CSF
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
9B4CSF
Granulocyte colony-stimulating factor, CSF3
Flow Cytometry,Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL