Please login to your online account to display your discounted pricing

Alfa Aesar™ G-A-Y

For cellular and molecular biology applications

Specifications

Product Type G-A-Y
Molecular Weight 309.32
Storage Requirements -30° to -10°C
Sequence Gly-Ala-Tyr
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66640MB

 
alfa aesar™
J66640MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66640MF

 
alfa aesar™
J66640MF
50mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type G-A-Y
Molecular Weight 309.32
Storage Requirements -30° to -10°C
Sequence Gly-Ala-Tyr
Format Solid
Regulatory Status RUO
Purification Method HPLC