Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fyn (p59 fyn) Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Fyn (p59fyn) Ab-1, Mouse Monoclonal Antibody.

$277.39 - $1,219.86

Specifications

Antigen Fyn (p59superscriptfynsuperscript) Ab-1
Clone 1S
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS339P

 
thermo scientific™ lab vision™
MS339P
Purified Ab with BSA and Azide 1mL Each for $1,135.08

MS339P0

 
thermo scientific™ lab vision™
MS339P0
Purified Ab with BSA and Azide 100µL Each for $277.39

MS339P1

 
thermo scientific™ lab vision™
MS339P1
Purified Ab with BSA and Azide 500µL Each for $745.06

MS339P1ABX

 
thermo scientific™ lab vision™
MS339P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $834.22

MS339PABX

 
thermo scientific™ lab vision™
MS339PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,219.86

MS339R7

 
thermo scientific™ lab vision™
MS339R7
Ready-to-use for IHC (UVLP) 7mL Each for $671.78
Description & Specifications

Specifications

Antigen Fyn (p59superscriptfynsuperscript) Ab-1
Clone 1S
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Immunogen Recombinant protein corresponding to aa 1-206 of murine p59superscriptfynsuperscriptf protein
Isotype IgG2a kappa

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Fyn (p59fyn) Ab-1, Mouse Monoclonal Antibody.

p59fyn is a membrane-associated non-receptor protein tyrosine kinase that belongs to the Src-family of kinases. The unique N-terminal domain of p59fyn interacts with the CD3 and eta chains of the TcR. p59fyn can bind other proteins (p82 and p116) through its SH2 and SH3 domains, which may act as substrates or regulators of fyn activity. p59ffyn is highly expressed in brain suggesting that it may have a role in the sensory nervous network and in myelination at early stages of CNS formation.

Host Species: Mouse

Clone: 1S

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 26-75

Immunogen: Recombinant protein corresponding to aa 1-206 of murine p59fyn protein

Molecular Weight: 59 kDa

Positive Control: Jurkat cells, Lymphoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO