Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Fyn (p59 fyn) Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Fyn (p59fyn) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS339P1ABX

 View more versions of this product

Catalog No. MS339P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Format Concentrated
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 100µL
Regulatory Status RUO
Clone 1S
Isotype IgG2a kappa
Immunogen Recombinant protein corresponding to aa 1-206 of murine p59superscriptfynsuperscriptf protein
Antigen Fyn (p59superscriptfynsuperscript) Ab-1
Formulation Ab without BSA/Azide (Frozen)
Classification Monoclonal
Concentration 1mg/mL
Type Primary
Research Discipline Signal Transduction
Conjugate Unlabeled
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Fyn (p59fyn) Ab-1, Mouse Monoclonal Antibody.

p59fyn is a membrane-associated non-receptor protein tyrosine kinase that belongs to the Src-family of kinases. The unique N-terminal domain of p59fyn interacts with the CD3 and eta chains of the TcR. p59fyn can bind other proteins (p82 and p116) through its SH2 and SH3 domains, which may act as substrates or regulators of fyn activity. p59ffyn is highly expressed in brain suggesting that it may have a role in the sensory nervous network and in myelination at early stages of CNS formation.

Host Species: Mouse

Clone: 1S

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 26-75

Immunogen: Recombinant protein corresponding to aa 1-206 of murine p59fyn protein

Molecular Weight: 59 kDa

Positive Control: Jurkat cells, Lymphoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Kinase Assay (Not verified)

Note:

USA: RUO; Int'l: RUO