missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human FYN A, GST Tag Recombinant Protein
Human FYN A, GST Tag Recombinant Protein
Supplier: Invitrogen™ PV6348
Description
Fyn has been shown to phosphorylate Dab1, an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2). The interaction of Fyn and Dab1 regulates the phosphorylation, trafficking, and processing of APP and apoEr2. Fyn expression has been shown to be significantly increased in Chronic Myelogenous Leukemia (CML). Knockdown of Fyn with shRNA slows leukemia cell growth, inhibits clonogenicity, and leads to increased sensitivity to imatinib.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
87.1 kDa | |
SLK, SYN, MGC45350 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Baculovirus | |
Liquid |
1mg | |
Human | |
GST Tag | |
Cytoplasmic Tyrosine Kinase | |
Recombinant |