missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

QSI Fused-Quartz Crucibles

Withstand temperatures up to 1250°C (2282°F)

Manufacturer:  QSI QTZ8072D

 View more versions of this product

Catalog No. 08-072D



Description

Description

Withstand temperatures up to 1250°C (2282°F)

  • Withstand severe temperature changes
  • Insoluble in water and acids except hydrofluoric and phosphoric acids
  • Covers sold separately

Specifications

Specifications

Clear
1250°C
30mL
3.8cm
High purity fused quartz
2mm
4.3cm
Quartz
SDS
Documents
Accessories