missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
QSI Fused-Quartz Crucibles
Withstand temperatures up to 1250°C (2282°F)
Manufacturer: QSI QTZ8072A
Description
Withstand temperatures up to 1250°C (2282°F)
- Withstand severe temperature changes
- Insoluble in water and acids except hydrofluoric and phosphoric acids
- Covers sold separately
Specifications
Clear | |
1250°C | |
10mL | |
3cm |
High purity fused quartz | |
2mm | |
2.9cm | |
Quartz |