missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fusaric acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC198962500

 View more versions of this product

Catalog No. AC198962500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Fusaric acid
Glass bottle
97°C to 101°C
536-69-6
C10H13NO2
15, 4343
DGMPVYSXXIOGJY-UHFFFAOYSA-N
5-butylpyridine-2-carboxylic acid
3442
179.22
99%
Authentic
White to Yellow
250mg
99%
MFCD00006298
2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, 5-butyl-pyridine-2-carboxylic acid, 5-butylpicolinic acid, 5-butylpyridine-3-carboxylic acid, 5-n-butylpyridine-2-carboxylic acid, fusaric acid, fusarinic acid, picolinic acid, 5-butyl, unii-jwj963070n
CCCCC1=CN=C(C=C1)C(=O)O
179.22
CHEBI:5199
Crystalline Powder

Chemical Identifiers

536-69-6
179.22
DGMPVYSXXIOGJY-UHFFFAOYSA-N
3442
5-butylpyridine-2-carboxylic acid
C10H13NO2
MFCD00006298
2-pyridinecarboxylic acid, 5-butyl, 5-butyl-2-pyridinecarboxylic acid, 5-butyl-pyridine-2-carboxylic acid, 5-butylpicolinic acid, 5-butylpyridine-3-carboxylic acid, 5-n-butylpyridine-2-carboxylic acid, fusaric acid, fusarinic acid, picolinic acid, 5-butyl, unii-jwj963070n
CHEBI:5199
CCCCC1=CN=C(C=C1)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 208-643-

SDS Documents