Please login to your online account to display your discounted pricing

Fusaric acid, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  198960010

 View more versions of this product

Catalog No. AC198960010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 179.22
Physical Form Crystalline Powder
Quantity 1g
Color Off-white to faint yellow
Merck Index 15, 4343
Melting Point 97° to 101°C
Infrared Spectrum Authentic
Synonym 5-Butyl-2-pyridinecarboxylic acid
CAS 536-69-6
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C10H13NO2
MDL Number MFCD00006298
Chemical Name or Material Fusaric acid
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.

Specifications

Chemical Name or Material Fusaric acid
CAS 536-69-6
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H13NO2
Formula Weight 179.22
MDL Number MFCD00006298
Synonym 5-Butyl-2-pyridinecarboxylic acid
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents