missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Furosemide, 97+%

Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

Manufacturer:  Alfa Aesar™ J6145714

 View more versions of this product

Catalog No. AAJ6145714



Specifications

Specifications

White
25g
C12H11ClN2O5S
14,4309
Soluble in acetone, DMF or methanol; Slightly soluble in water
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
330.739
CHEBI:47426
Crystalline Powder
206°C to 209°C
54-31-9
MFCD00010549
aisemide, beronald, desdemin, errolon, frusemide, furanthril, furosemid, furosemide, fusid, lasix
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
3440
330.74
≥97%

Chemical Identifiers

54-31-9
330.739
ZZUFCTLCJUWOSV-UHFFFAOYSA-N
3440
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
C12H11ClN2O5S
MFCD00010549
aisemide, beronald, desdemin, errolon, frusemide, furanthril, furosemid, furosemide, fusid, lasix
CHEBI:47426
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H360

EINECSNumber : 200-203-6

RTECSNumber : CB2625000

TSCA : No

Recommended Storage : Ambient temperatures

Documents