Learn More
Furosemide, 97+%, Thermo Scientific Chemicals
Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter
Supplier: Thermo Scientific Chemicals J6145709
Description
Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Furosemide | |
206°C to 209°C | |
C12H11ClN2O5S | |
10 g | |
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin | |
ZZUFCTLCJUWOSV-UHFFFAOYSA-N | |
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid | |
3440 | |
330.74 | |
Crystalline Powder |
54-31-9 | |
White | |
MFCD00010549 | |
14,4309 | |
Soluble in acetone, DMF or methanol; Slightly soluble in water | |
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl | |
330.739 | |
CHEBI:47426 | |
≥97% |
Chemical Identifiers
54-31-9 | |
330.739 | |
ZZUFCTLCJUWOSV-UHFFFAOYSA-N | |
3440 | |
4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid |
C12H11ClN2O5S | |
MFCD00010549 | |
furosemide, frusemide, lasix, furosemid, furanthril, errolon, fusid, aisemide, beronald, desdemin | |
CHEBI:47426 | |
C1=COC(=C1)CNC2=CC(=C(C=C2C(=O)O)S(=O)(=O)N)Cl |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360
EINECSNumber : 200-203-6
RTECSNumber : CB2625000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only