Please login to your online account to display your discounted pricing

Alfa Aesar™ (Furfurylthio)acetone, 96%

$83.18 - $83.18

Chemical Identifiers

Chemical Name or Material (Furfurylthio)acetone
CAS 58066-86-7
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD02683098
Synonym 1-(Furfurylthio)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2272406

  SDS
alfa aesar™
B2272406
5g Each for $83.18
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material (Furfurylthio)acetone
Assay 96%
Molecular Formula C8H10O2S
MDL Number MFCD02683098
CAS 58066-86-7
Synonym 1-(Furfurylthio)-2-propanone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.