missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfurylamine, TCI America™

Manufacturer:  TCI America F009125ML

 View more versions of this product

Catalog No. F009125ML



Specifications

Specifications

Furfurylamine
Yellow
25mL
C5H7NO
2526
DDRPCXLAQZKBJP-UHFFFAOYSA-N
furan-2-ylmethanamine
3438
Clear Liquid at 20°C
146°C
-70°C
617-89-0
MFCD00003258
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
C1=COC(=C1)CN
97.117
97.12
≥98.0% (GC,T)

Chemical Identifiers

617-89-0
97.117
DDRPCXLAQZKBJP-UHFFFAOYSA-N
3438
C1=COC(=C1)CN
C5H7NO
MFCD00003258
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
furan-2-ylmethanamine
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1264

RTECSNumber : LU9275000

TSCA : Yes

Documents