Learn More
Furfurylamine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119802500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Furfurylamine | |
617-89-0 | |
Brown-Yellow to Colorless | |
145°C to 146°C (761.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4890 to 1.491 | |
250 mL | |
1.09 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
C1=COC(=C1)CN | |
97.12 | |
1.6 mPa.s (20°C) | |
99+% |
99+% | |
-70°C | |
1.0900g/mL | |
37°C | |
99+% | |
C5H7NO | |
MFCD00003258 | |
18, 584 | |
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl | |
DDRPCXLAQZKBJP-UHFFFAOYSA-N | |
furan-2-ylmethanamine | |
3438 | |
97.12 | |
Liquid |
Chemical Identifiers
617-89-0 | |
97.12 | |
DDRPCXLAQZKBJP-UHFFFAOYSA-N | |
3438 | |
C1=COC(=C1)CN |
C5H7NO | |
MFCD00003258 | |
furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl | |
furan-2-ylmethanamine |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and
GHS Signal Word: Danger
EINECSNumber : 210-536-9
RTECSNumber : LU9275000
TSCA : TSCA
RUO – Research Use Only