Please login to your online account to display your discounted pricing

Alfa Aesar™ Furfuryl methyl sulfide, 98%

$85.58 - $299.85

Chemical Identifiers

Chemical Name or Material Furfuryl methyl sulfide
CAS 1438-91-1
Molecular Formula C6H8OS
Formula Weight 128.19
MDL Number MFCD00009604
Beilstein 107109
Synonym 2-(Methylthiomethyl)furan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2169114

  SDS
alfa aesar™
B2169114
25g Each for $85.58

AAB2169122

  SDS
alfa aesar™
B2169122
100g Each for $299.85
Description & Specifications

Specifications

Formula Weight 128.19
Flash Point 63°C (145°F)
Chemical Name or Material Furfuryl methyl sulfide
TSCA TSCA
Assay 98%
Molecular Formula C6H8OS
MDL Number MFCD00009604
CAS 1438-91-1
Beilstein 107109
Synonym 2-(Methylthiomethyl)furan
Health Hazard 1 Warning
Boiling Point 170° to 173°C
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Quantity 25g
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.