missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl Methacrylate (stabilized with MEHQ), TCI America™

$64.17 - $220.00

Chemical Identifiers

CAS 3454-28-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00046113
InChI Key DWXAVNJYFLGAEF-UHFFFAOYSA-N
Synonym Methacrylic Acid Furfuryl Ester
PubChem CID 76993
IUPAC Name furan-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13355G View Documents TCI America
M13355G
5g Each for $64.17
 
M133525G View Documents TCI America
M133525G
25g Each for $220.00
 
Specifications

Chemical Identifiers

3454-28-2
166.176
DWXAVNJYFLGAEF-UHFFFAOYSA-N
76993
CC(=C)C(=O)OCC1=CC=CO1
C9H10O3
MFCD00046113
Methacrylic Acid Furfuryl Ester
furan-2-ylmethyl 2-methylprop-2-enoate

Specifications

82°C
5g
C9H10O3
Methacrylic Acid Furfuryl Ester
CC(=C)C(=O)OCC1=CC=CO1
166.176
166.18
≥95.0% (GC)
Yellow
3454-28-2
MFCD00046113
DWXAVNJYFLGAEF-UHFFFAOYSA-N
furan-2-ylmethyl 2-methylprop-2-enoate
76993
Clear Liquid at 20°C
Furfuryl Methacrylate (stabilized with MEHQ)
Safety and Handling

Safety and Handling

TSCA : Yes